Thermo Scientific Chemicals

Dimethylphosphoramidous Dichloride, 97%, Thermo Scientific Chemicals

Catalog number: 383350050
5 mL, Glass bottle, Each
Thermo Scientific Chemicals

Dimethylphosphoramidous Dichloride, 97%, Thermo Scientific Chemicals

Catalog number: 383350050
5 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 383350050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
683-85-2
IUPAC Name
(dichlorophosphanyl)dimethylamine
Molecular Formula
C2H6Cl2NP
InChI Key
XPWWDZRSNFSLRQ-UHFFFAOYSA-N
SMILES
CN(C)P(Cl)Cl
Infrared spectrum
Conforms
Appearance (Color)
Colorless to light yellow
NMR
>=96.0 %
Appearance (Form)
Liquid
Refractive index
1.5040 to 1.5060 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text