Thermo Scientific Chemicals

Bis(2-diphenylphosphinophenyl)ether, 99%, Thermo Scientific Chemicals

Catalog number: 383370250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Bis(2-diphenylphosphinophenyl)ether, 99%, Thermo Scientific Chemicals

Catalog number: 383370250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 383370250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
166330-10-5
IUPAC Name
{2-[2-(diphenylphosphanyl)phenoxy]phenyl}diphenylphosphane
Molecular Formula
C36H28OP2
InChI Key
RYXZOQOZERSHHQ-UHFFFAOYSA-N
SMILES
O(C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
HPLC
>=98.5 %
Appearance (Form)
Crystalline powder or crystals
Melting point
184°C to 187°C
Appearance (Color)
White to off-white
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text