Diisopropylphosphine, 10 wt.% solution in hexanes, Thermo Scientific Chemicals
Diisopropylphosphine, 10 wt.% solution in hexanes, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diisopropylphosphine, 10 wt.% solution in hexanes, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 383400025
Price (USD)/ Each
89.20
-
Add to cart
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
89.20
Add to cart
Diisopropylphosphine, 10 wt.% solution in hexanes, Thermo Scientific Chemicals
Catalog number383400025
Price (USD)/ Each
89.20
-
Add to cart
Chemical Identifiers
IUPAC Namebis(propan-2-yl)phosphane
Molecular FormulaC6H15P
InChI KeyWDIIYWASEVHBBT-UHFFFAOYSA-N
SMILESCC(C)PC(C)C
Molecular Weight (g/mol)118.16
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration9 to 11 wt%
Appearance (Form)Solution
Appearance (Color)Clear colorless
NMR (31P-NMR)Consistent with structure
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only