Thermo Scientific Chemicals

Diisopropylphosphine, 10 wt.% solution in hexanes, Thermo Scientific Chemicals

Catalog number: 383400100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Diisopropylphosphine, 10 wt.% solution in hexanes, Thermo Scientific Chemicals

Catalog number: 383400100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 383400100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20491-53-6, 110-54-3
Appearance (Color)
Clear colorless
NMR (31P-NMR)
Consistent with structure
Concentration
9 to 11 wt%
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text