Chemical Identifiers
CAS11136-36-0
IUPAC Nametitanium(4+) bis(pentamethylcyclopenta-1,3-dien-1-ide) dichloride
Molecular FormulaC20H30Cl2Ti
InChI KeyZBFBXTFQCKIUHU-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Ti+4].CC1=[C-]C(C)(C)C(C)=C1C.CC1=[C-]C(C)(C)C(C)=C1C
View more