Thermo Scientific Chemicals

Ethyl isobutyrylacetate, 95%, Thermo Scientific Chemicals

Catalog number: 383440100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl isobutyrylacetate, 95%, Thermo Scientific Chemicals

Catalog number: 383440100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 383440100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7152-15-0
IUPAC Name
ethyl 4-methyl-3-oxopentanoate
Molecular Formula
C8H14O3
InChI Key
XCLDSQRVMMXWMS-UHFFFAOYSA-N
SMILES
CCOC(=O)CC(=O)C(C)C
Appearance (Form)
Liquid
Refractive index
1.4240 to 1.4280 (20°C, 589 nm)
GC
>=94 % (Mixture of tautomers)
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text