Thermo Scientific Chemicals

Gadolinium(III) chloride, 99.9%, (trace metal basis), anhydrous, Thermo Scientific Chemicals

Catalog number: 383560250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Gadolinium(III) chloride, 99.9%, (trace metal basis), anhydrous, Thermo Scientific Chemicals

Catalog number: 383560250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 383560250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10138-52-0
IUPAC Name
gadolinium(3+) trichloride
Molecular Formula
Cl3Gd
InChI Key
MEANOSLIBWSCIT-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Gd+3]
Appearance (Color)
White to off-white
Total trace metal impurities
=<1000 ppm
Titration Complexometric
58.2 to 61.1 % (Gd)
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text