cis-2-Pentene
cis-2-Pentene
cis-2-Pentene
Thermo Scientific Chemicals

cis-2-Pentene

CAS: 627-20-3 | C5H10 | 70.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
10 g
Packaging:
Glass bottle
Catalog number 383590100
Price (USD)
189.65
Special offer
Online exclusive
Ends: 26-Sep-2025
234.00
Save 44.35 (19%)
Each
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
189.65
Special offer
Online exclusive
Ends: 26-Sep-2025
234.00
Save 44.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS627-20-3
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=95.0 %
Appearance (Form)Liquid
Refractive index1.3810 to 1.3830 (20°C, 589 nm)
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link