Lanthanum(III) Isopropoxide, 98%, Thermo Scientific Chemicals
Lanthanum(III) Isopropoxide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lanthanum(III) Isopropoxide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 383640050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS19446-52-7
IUPAC Namelanthanum(3+) tris(propan-2-olate)
Molecular FormulaC9H21LaO3
InChI KeySORGMJIXNUWMMR-UHFFFAOYSA-N
SMILES[La+3].CC(C)[O-].CC(C)[O-].CC(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
Appearance (Form)Powder
Titration Complexometric>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only