5-Hexynoic acid, 97%, Thermo Scientific Chemicals
5-Hexynoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Hexynoic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 383700010
Price (USD)/ Each
56.50
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
56.50
Add to cart
5-Hexynoic acid, 97%, Thermo Scientific Chemicals
Catalog number383700010
Price (USD)/ Each
56.50
-
Add to cart
Chemical Identifiers
CAS53293-00-8
IUPAC Namehex-5-ynoate
Molecular FormulaC6H7O2
InChI KeyVPFMEXRVUOPYRG-UHFFFAOYSA-M
SMILES[O-]C(=O)CCCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear yellow
Titration with NaOH>=96.0 %
Infrared spectrumConforms
Refractive index1.4480 to 1.4500 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only