Thermo Scientific Chemicals

Dichloromethane 99+%, stabilized with amylene, pure, Thermo Scientific Chemicals

Catalog number: 383780250
25 L, Each
Thermo Scientific Chemicals

Dichloromethane 99+%, stabilized with amylene, pure, Thermo Scientific Chemicals

Catalog number: 383780250
25 L, Each
Quantity
Catalog number: 383780250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-09-2
IUPAC Name
dichloromethane
Molecular Formula
CH2Cl2
InChI Key
YMWUJEATGCHHMB-UHFFFAOYSA-N
SMILES
ClCCl
Appearance (Form)
Liquid
Residue after evaporation
=<0.05 %
GC
>=99.0 %
Stabilizer
Approx. 100 ppm Amylene
Water
=<0.05 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text