Thermo Scientific Chemicals

n-Heptane, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 383790050
5 L, Each
Thermo Scientific Chemicals

n-Heptane, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 383790050
5 L, Each
Quantity
Catalog number: 383790050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
142-82-5
IUPAC Name
heptane
Molecular Formula
C7H16
InChI Key
IMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILES
CCCCCCC
Appearance (Form)
Liquid
GC
>=99.0 %
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Residue after evaporation
=<20 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text