Thermo Scientific Chemicals

Isohexane, for HPLC, contains <5% n-Hexane, Thermo Scientific Chemicals

Catalog number: 383810025
2.5 L, Each
Thermo Scientific Chemicals

Isohexane, for HPLC, contains <5% n-Hexane, Thermo Scientific Chemicals

Catalog number: 383810025
2.5 L, Each
Quantity
Catalog number: 383810025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
73513-42-5
IUPAC Name
2-methylpentane
Molecular Formula
C6H14
InChI Key
AFABGHUZZDYHJO-UHFFFAOYSA-N
SMILES
CCCC(C)C
Appearance (Form)
Liquid
UV
at 200 nm A: =<0.3
UV
at 220 nm A: =<0.07
UV
at 240 nm A: =<0.02
UV
at 260 nm A: =<0.005

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text