1x1 image pixel for data collection
Buffer solution pH 4, Phthalate buffer, traceable to NIST, ready to use
Buffer solution pH 4, Phthalate buffer, traceable to NIST, ready to use
Buffer solution pH 4, Phthalate buffer, traceable to NIST, ready to use
Buffer solution pH 4, Phthalate buffer, traceable to NIST, ready to use
Thermo Scientific Chemicals

Buffer solution pH 4, Phthalate buffer, traceable to NIST, ready to use

CAS: 877-24-7 | C8H5KO4
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number 383835000
Price (USD)
21.20
Each
In stock
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)
21.20
Each
Add to cart
Chemical Identifiers
CAS877-24-7
IUPAC Namepotassium 2-carboxybenzoate
Molecular FormulaC8H5KO4
InChI KeyIWZKICVEHNUQTL-UHFFFAOYSA-M
SMILES[K+].OC(=O)C1=CC=CC=C1C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
pH3.98 to 4.02 (at 20°C)
Appearance (Color)Clear colorless
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link