Buffer solution pH 7, Phosphate buffer, traceable to NIST, ready to use
Buffer solution pH 7, Phosphate buffer, traceable to NIST, ready to use
Buffer solution pH 7, Phosphate buffer, traceable to NIST, ready to use
Buffer solution pH 7, Phosphate buffer, traceable to NIST, ready to use
Thermo Scientific Chemicals

Buffer solution pH 7, Phosphate buffer, traceable to NIST, ready to use

CAS: 7558-79-4
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Catalog number 383850010
Price (USD)
56.90
Each
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)
56.90
Each
Add to cart
Chemical Identifiers
CAS7558-79-4
IUPAC Namedisodium hydrogen phosphate
Molecular FormulaHNa2O4P
InChI KeyBNIILDVGGAEEIG-UHFFFAOYSA-L
SMILES[Na+].[Na+].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Clear colorless
pH6.98 to 7.02 (at 20°C)
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection