Thermo Scientific Chemicals

Buffer solution pH 9.2, Borate buffer, traceable to NIST, ready to use, Thermo Scientific Chemicals

Catalog number: 383875000
500 mL, Each
Thermo Scientific Chemicals

Buffer solution pH 9.2, Borate buffer, traceable to NIST, ready to use, Thermo Scientific Chemicals

Catalog number: 383875000
500 mL, Each
Quantity
Catalog number: 383875000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1330-43-4, 7732-18-5, 7647-14-5
Appearance (Color)
Clear colorless
pH
9.18 to 9.22 (at 20°C)
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text