Thermo Scientific Chemicals

n-Pentane, for analysis, Thermo Scientific Chemicals

Catalog number: 383900100
10 L, Metal Drum, Each
Thermo Scientific Chemicals

n-Pentane, for analysis, Thermo Scientific Chemicals

Catalog number: 383900100
10 L, Metal Drum, Each
Quantity
Packaging
Catalog number: 383900100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
109-66-0
IUPAC Name
pentane
Molecular Formula
C5H12
InChI Key
OFBQJSOFQDEBGM-UHFFFAOYSA-N
SMILES
CCCCC
Residue after evaporation
=<0.002 %
Magnesium (Mg)
=<0.1 ppm
Total phosphorus (P)
=<0.5 ppm
Color scale
=<10 APHA
Copper (Cu)
=<0.05 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text