Thermo Scientific Chemicals

Isopropanol, 99.5%, Thermo Scientific Chemicals

Catalog number: 383910010
1 L, Each
Thermo Scientific Chemicals

Isopropanol, 99.5%, Thermo Scientific Chemicals

Catalog number: 383910010
1 L, Each
Quantity
Catalog number: 383910010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67-63-0
IUPAC Name
propan-2-ol
Molecular Formula
C3H8O
InChI Key
KFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILES
CC(C)O
Appearance (Form)
Clear liquid
GC
>=99.5 %
Acidity/alkalinity
=<0.0001 meq/g
UV
at 220 nm A: =<0.2
UV
at 240 nm A: =<0.05

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text