Isopropanol, 99.5%, for HPLC
Isopropanol, 99.5%, for HPLC
Isopropanol, 99.5%, for HPLC
Isopropanol, 99.5%, for HPLC
Thermo Scientific Chemicals

Isopropanol, 99.5%, for HPLC

CAS: 67-63-0 | C3H8O | 60.096 g/mol
Catalog NumberQuantity
3839100252.5 L
Catalog number 383910025
Price (JPY)
-
Request A Quote
Quantity:
2.5 L
Request bulk or custom format
Chemical Identifiers
CAS67-63-0
IUPAC Namepropan-2-ol
Molecular FormulaC3H8O
InChI KeyKFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILESCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 230 nm A: =<0.1
UVat 250 nm A: =<0.01
UVat 270 nm A: =<0.005
Residue after evaporation=<2 ppm
UVat 260 nm A: =<0.005
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link