Isopropanol, 99.5%, for HPLC gradient grade
Isopropanol, 99.5%, for HPLC gradient grade
Isopropanol, 99.5%, for HPLC gradient grade
Isopropanol, 99.5%, for HPLC gradient grade
Thermo Scientific Chemicals

Isopropanol, 99.5%, for HPLC gradient grade

CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number 383920010
Price (USD)
63.65
Online Exclusive
71.00
Save 7.35 (10%)
Each
In stock
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)
63.65
Online Exclusive
71.00
Save 7.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS67-63-0
IUPAC Namepropan-2-ol
Molecular FormulaC3H8O
InChI KeyKFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILESCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Acidity/alkalinity=<0.0001 meq/g
Filtered to 0.2 micrometerFiltered
GC>=99.4 %
UVat 205 nm Cut off
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link