Isopropanol, 99.5%, for HPLC gradient grade, Thermo Scientific Chemicals
Isopropanol, 99.5%, for HPLC gradient grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isopropanol, 99.5%, for HPLC gradient grade, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number 383920025
Price (USD)/ Each
113.65
Online exclusive
126.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (USD)/ Each
113.65
Online exclusive
126.00 
Save 12.35 (10%)
Add to cart
Isopropanol, 99.5%, for HPLC gradient grade, Thermo Scientific Chemicals
Catalog number383920025
Price (USD)/ Each
113.65
Online exclusive
126.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS67-63-0
IUPAC Namepropan-2-ol
Molecular FormulaC3H8O
InChI KeyKFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILESCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Water=<0.1 %
Residue after evaporation=<2 ppm
UVat 240 nm A: =<0.05
UV1 cm cell vs HPLC-grade water
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only