Xylenes, 98+%, for analysis, mixture of isomers , Thermo Scientific Chemicals
Xylenes, 98+%, for analysis, mixture of isomers , Thermo Scientific Chemicals
Thermo Scientific Chemicals

Xylenes, 98+%, for analysis, mixture of isomers , Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 383940010
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
61.50 
Save 8.85 (14%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
61.50 
Save 8.85 (14%)
Add to cart
Xylenes, 98+%, for analysis, mixture of isomers , Thermo Scientific Chemicals
Catalog number383940010
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
61.50 
Save 8.85 (14%)
-
Add to cart
Chemical Identifiers
CAS1330-20-7
IUPAC Name1,3-xylene
Molecular FormulaC8H10
InChI KeyIVSZLXZYQVIEFR-UHFFFAOYSA-N
SMILESCC1=CC(C)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Water=<0.05 % (K.F.)
Copper (Cu)=<0.05 ppm
Lead (Pb)=<0.05 ppm
Potassium (K)=<0.5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only