Titanium(IV) oxide, Aeroxide(R) P25, Thermo Scientific Chemicals
Titanium(IV) oxide, Aeroxide(R) P25, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Titanium(IV) oxide, Aeroxide(R) P25, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
250 g
Packaging:
View all
Plastic drum
Catalog number 384290010
Price (USD)/ Each
264.65
Online exclusive
294.00 
Save 29.35 (10%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
264.65
Online exclusive
294.00 
Save 29.35 (10%)
Add to cart
Titanium(IV) oxide, Aeroxide(R) P25, Thermo Scientific Chemicals
Catalog number384290010
Price (USD)/ Each
264.65
Online exclusive
294.00 
Save 29.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13463-67-7
IUPAC Namedioxotitanium
Molecular FormulaO2Ti
InChI KeyGWEVSGVZZGPLCZ-UHFFFAOYSA-N
SMILESO=[Ti]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
pH3.5 to 4.5 (4 % dispersion)
Appearance (Color)White
Specific surface area35 to 65 m²/g
Acidity (HCl)=<0.3 %
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only