Titanium(IV) oxide, Aeroxide™ P25
Titanium(IV) oxide, Aeroxide™ P25
Titanium(IV) oxide, Aeroxide™ P25
Titanium(IV) oxide, Aeroxide™ P25
Titanium(IV) oxide, Aeroxide™ P25
Thermo Scientific Chemicals

Titanium(IV) oxide, Aeroxide™ P25

CAS: 13463-67-7 | O2Ti | 79.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number 384290500
Price (USD)
70.65
Online Exclusive
78.30
Save 7.65 (10%)
Each
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
70.65
Online Exclusive
78.30
Save 7.65 (10%)
Each
Add to cart
Chemical Identifiers
CAS13463-67-7
IUPAC Namedioxotitanium
Molecular FormulaO2Ti
InChI KeyGWEVSGVZZGPLCZ-UHFFFAOYSA-N
SMILESO=[Ti]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Specific surface area35 to 65 m²/g
Acidity (HCl)=<0.3 %
Appearance (Form)Powder
pH3.5 to 4.5 (4 % dispersion)
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Share catalog number, name or link