Thermo Scientific Chemicals

Titanium(IV) oxide, Aeroxide(R) P25, Thermo Scientific Chemicals

Catalog number: 384292500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Titanium(IV) oxide, Aeroxide(R) P25, Thermo Scientific Chemicals

Catalog number: 384292500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 384292500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13463-67-7
IUPAC Name
dioxotitanium
Molecular Formula
O2Ti
InChI Key
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
SMILES
O=[Ti]=O
Appearance (Color)
White
Specific surface area
35 to 65 m²/g
Acidity (HCl)
=<0.3 %
Assay
>=99.5 % (on ignited substance) Typical
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text