1(2H)-Phthalazinone, 98+%, Thermo Scientific Chemicals
1(2H)-Phthalazinone, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1(2H)-Phthalazinone, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 384575000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name1,2-dihydrophthalazin-1-one
Molecular FormulaC8H6N2O
InChI KeyIJAPPYDYQCXOEF-UHFFFAOYSA-N
SMILESO=C1NN=CC2=CC=CC=C12
Molecular Weight (g/mol)146.15
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light yellow
Melting point184°C to 187°C
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only