10-Bromo-1-decanol, 95%, Thermo Scientific Chemicals
10-Bromo-1-decanol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

10-Bromo-1-decanol, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 384590250
Price (USD)/ Each
343.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
343.00
Add to cart
10-Bromo-1-decanol, 95%, Thermo Scientific Chemicals
Catalog number384590250
Price (USD)/ Each
343.00
-
Add to cart
Chemical Identifiers
IUPAC Name10-bromodecan-1-ol
Molecular FormulaC10H21BrO
InChI KeyLGZMUUBPTDRQQM-UHFFFAOYSA-N
SMILESOCCCCCCCCCCBr
Molecular Weight (g/mol)237.18
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4780 to 1.4800 (20°C, 589 nm)
Appearance (Color)Clear yellow
GC>=94 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only