Thermo Scientific Chemicals

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylic Acid Tert-Butyl Ester, 97%, Thermo Scientific Chemicals

Catalog number: 384640050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylic Acid Tert-Butyl Ester, 97%, Thermo Scientific Chemicals

Catalog number: 384640050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 384640050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
552846-17-0
IUPAC Name
tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate
Molecular Formula
C14H23BN2O4
InChI Key
IPISOFJLWYBCAV-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)N1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
Appearance (Form)
Crystalline powder
Melting point
82°C to 89°C
Infrared spectrum
Conforms
HPLC
>=96.0 %
Appearance (Color)
White to beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text