1-Octanesulfonic acid, sodium salt, HPLC grade, Thermo Scientific Chemicals
1-Octanesulfonic acid, sodium salt, HPLC grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Octanesulfonic acid, sodium salt, HPLC grade, Thermo Scientific Chemicals

1-Octanesulfonic acid, sodium salt, HPLC grade, Acros Organics, Quantity: 25g, White, Melting Point: >300.0 deg.C, Molecular Weight: 216.28, Percent Purity: >-98.0%
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Catalog number 384770050
Price (USD)/ Each
66.65
Online exclusive
78.30 
Save 11.65 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
66.65
Online exclusive
78.30 
Save 11.65 (15%)
Add to cart
1-Octanesulfonic acid, sodium salt, HPLC grade, Thermo Scientific Chemicals
Catalog number384770050
Price (USD)/ Each
66.65
Online exclusive
78.30 
Save 11.65 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namesodium octane-1-sulfonate
Molecular FormulaC8H17NaO3S
InChI KeyHRQDCDQDOPSGBR-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCS([O-])(=O)=O
Molecular Weight (g/mol)216.27
View more
SpecificationsSpecification SheetSpecification Sheet
Titration after Ion exchange>=98.0 %
UVat 200 nm A: =<0.3
UVat 220 nm A: =<0.1
UVat 240 nm A: =<0.05
UVat 260 nm A: =<0.04
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography
RUO – Research Use Only