Thermo Scientific Chemicals

1-Octanesulfonic acid, sodium salt, HPLC grade, Thermo Scientific Chemicals

Catalog number: 384770250
25 g, Each
Thermo Scientific Chemicals

1-Octanesulfonic acid, sodium salt, HPLC grade, Thermo Scientific Chemicals

Catalog number: 384770250
25 g, Each
Quantity
Catalog number: 384770250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5324-84-5
IUPAC Name
sodium octane-1-sulfonate
Molecular Formula
C8H17NaO3S
InChI Key
HRQDCDQDOPSGBR-UHFFFAOYSA-M
SMILES
[Na+].CCCCCCCCS([O-])(=O)=O
Titration after Ion exchange
>=98.0 %
UV
at 200 nm A: =<0.3
UV
at 220 nm A: =<0.1
UV
at 240 nm A: =<0.05
UV
at 260 nm A: =<0.04

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text