Thermo Scientific Chemicals

1-Isopropylpiperazine, 98+%, Thermo Scientific Chemicals

Catalog number: 384810050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Isopropylpiperazine, 98+%, Thermo Scientific Chemicals

Catalog number: 384810050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 384810050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4318-42-7
IUPAC Name
1-(propan-2-yl)piperazine
Molecular Formula
C7H16N2
InChI Key
WHKWMTXTYKVFLK-UHFFFAOYSA-N
SMILES
CC(C)N1CCNCC1
GC
>=98.0 %
Appearance (Form)
Liquid
Refractive index
1.4700 to 1.4720 (20°C, 589 nm)
Appearance (Color)
Clear light yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text