Thermo Scientific Chemicals

N-BOC-4-Piperidinecarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 384870050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

N-BOC-4-Piperidinecarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 384870050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 384870050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
84358-13-4
IUPAC Name
1-[(tert-butoxy)carbonyl]piperidine-4-carboxylic acid
Molecular Formula
C11H19NO4
InChI Key
JWOHBPPVVDQMKB-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)N1CCC(CC1)C(O)=O
Appearance (Color)
White
Infrared spectrum
Conforms
Titration with NaOH
>=97.5 %
Appearance (Form)
Crystalline powder
Melting point
149°C to 153°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text