Thermo Scientific Chemicals

n-Octadecyl sulfate, sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: 384890250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

n-Octadecyl sulfate, sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: 384890250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 384890250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1120-04-3
IUPAC Name
sodium octadecyl sulfate
Molecular Formula
C18H37NaO4S
InChI Key
NWZBFJYXRGSRGD-UHFFFAOYSA-M
SMILES
[Na+].CCCCCCCCCCCCCCCCCCOS([O-])(=O)=O
Appearance (Color)
White to light yellow to light beige
Infrared spectrum
Conforms
Loss on drying
=<7 %
Appearance (Form)
Powder
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text