Thermo Scientific Chemicals

1-Octadecanesulfonic acid, sodium salt, 90%, Thermo Scientific Chemicals

Catalog number: 384910050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Octadecanesulfonic acid, sodium salt, 90%, Thermo Scientific Chemicals

Catalog number: 384910050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 384910050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13893-34-0
IUPAC Name
octadecane-1-sulfonic acid sodium
Molecular Formula
C18H38NaO3S
InChI Key
NIMUYFTYMXTPAV-UHFFFAOYSA-N
SMILES
[Na].CCCCCCCCCCCCCCCCCCS(O)(=O)=O
Appearance (Form)
Powder
Infrared spectrum
Conforms
Elemental Analysis
>=88 % (C, H)
Appearance (Color)
Off-white
Loss on drying
=<5 % (1 g, 105°C, 1 h, 760 mmHg)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text