Chemical Identifiers
CAS2001-95-8
IUPAC Name(3S,6S,9R,12R,15S,18S,21R,24R,27S,30S,36R)-6,18,30-trimethyl-3,9,12,15,21,24,27,33,36-nonakis(propan-2-yl)-1,7,13,19,25,31-hexaoxa-4,10,16,22,28,34-hexaazacyclohexatriacontane-2,5,8,11,14,17,20,23,26,29,32,35-dodecone
Molecular FormulaC54H90N6O18
InChI KeyFCFNRCROJUBPLU-UYBNATROSA-N
SMILESCC(C)[C@@H]1NC(=O)[C@H](C)OC(=O)C(NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC1=O)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C
View more