Osmium 99.9%, Thermo Scientific Chemicals
Osmium 99.9%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Osmium 99.9%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 385160010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-04-2
IUPAC Nameosmium
Molecular FormulaOs
InChI KeySYQBFIAQOQZEGI-UHFFFAOYSA-N
SMILES[Os]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Total trace metal impurities=<0.1 %
Particle Size/Sieve analysis=<100 mesh
Appearance (Color)Blue-grey
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only