4-Aminomethyltetrahydropyran, 97%, Thermo Scientific Chemicals
4-Aminomethyltetrahydropyran, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Aminomethyltetrahydropyran, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 385210010
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
IUPAC Name(oxan-4-yl)methanaminium
Molecular FormulaC6H14NO
InChI KeyIPBPLHNLRKRLPJ-UHFFFAOYSA-O
SMILES[NH3+]CC1CCOCC1
Molecular Weight (g/mol)116.18
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4640 to 1.4670 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only