Thermo Scientific Chemicals

4-Aminomethyltetrahydropyran, 97%, Thermo Scientific Chemicals

Catalog number: 385210250
25 g, Each
Thermo Scientific Chemicals

4-Aminomethyltetrahydropyran, 97%, Thermo Scientific Chemicals

Catalog number: 385210250
25 g, Each
Quantity
Catalog number: 385210250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
130290-79-8
IUPAC Name
(oxan-4-yl)methanaminium
Molecular Formula
C6H14NO
InChI Key
IPBPLHNLRKRLPJ-UHFFFAOYSA-O
SMILES
[NH3+]CC1CCOCC1
Appearance (Color)
Colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.4640 to 1.4670 (20°C, 589 nm)
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text