Thermo Scientific Chemicals

(Trimethylsilyl)diazomethane, 2M solution in hexanes, ACROS Organics™

Catalog number: 385335000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

(Trimethylsilyl)diazomethane, 2M solution in hexanes, ACROS Organics™

Catalog number: 385335000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 385335000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18107-18-1
IUPAC Name
(diazomethyl)trimethylsilane
Molecular Formula
C4H10N2Si
InChI Key
ONDSBJMLAHVLMI-UHFFFAOYSA-N
SMILES
C[Si](C)(C)C=[N+]=[N-]
Appearance (Form)
Solution
Appearance (Color)
Clear yellow
Strength
1.8 to 2.4 M

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text