Thermo Scientific Chemicals

Calcium sulfate hemihydrate, 97+%, pure, Thermo Scientific Chemicals

Catalog number: 385350025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Calcium sulfate hemihydrate, 97+%, pure, Thermo Scientific Chemicals

Catalog number: 385350025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 385350025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10034-76-1
IUPAC Name
dicalcium hydrate disulfate
Molecular Formula
Ca2H2O9S2
InChI Key
ZOMBKNNSYQHRCA-UHFFFAOYSA-J
SMILES
O.[Ca++].[Ca++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Loss on ignition
4.5 % to 8.0 % (500°C, 2 h)
Appearance (Color)
White to off-white
Identification
Confirmed
Appearance (Form)
Powder
Titration Complexometric
>=97.0 % (On dry substance)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text