Calcium sulfate hemihydrate, 97+%, pure
Calcium sulfate hemihydrate, 97+%, pure
Calcium sulfate hemihydrate, 97+%, pure
Calcium sulfate hemihydrate, 97+%, pure
Thermo Scientific Chemicals

Calcium sulfate hemihydrate, 97+%, pure

CAS: 10034-76-1 | 0·5 H2O | 145.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 385351000
Price (EUR)
19,81
Online Exclusive
23,30
Save 3,49 (15%)
Each
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
19,81
Online Exclusive
23,30
Save 3,49 (15%)
Each
Add to cart
Chemical Identifiers
CAS10034-76-1
IUPAC Namedicalcium hydrate disulfate
Molecular FormulaCa2H2O9S2
InChI KeyZOMBKNNSYQHRCA-UHFFFAOYSA-J
SMILESO.[Ca++].[Ca++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on ignition4.5 % to 8.0 % (500°C, 2 h)
Appearance (Color)White to off-white
IdentificationConfirmed
Appearance (Form)Powder
Titration Complexometric>=97.0 % (On dry substance)
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0470924Certificate of AnalysisMay 17, 2025385350025, 385355000, 385351000
A0464699Certificate of AnalysisAug 08, 2024385350025, 385355000, 385351000
A0454834Certificate of AnalysisAug 25, 2023385350025, 385355000, 385351000
A0398840Certificate of AnalysisDec 14, 2022385350025, 385355000, 385351000
A0410584Certificate of AnalysisDec 13, 2022385350025, 385355000, 385351000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection