Thermo Scientific Chemicals

Ethyl myristate, 98+%, Thermo Scientific Chemicals

Catalog number: 385541000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl myristate, 98+%, Thermo Scientific Chemicals

Catalog number: 385541000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 385541000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
124-06-1
IUPAC Name
ethyl tetradecanoate
Molecular Formula
C16H32O2
InChI Key
MMKRHZKQPFCLLS-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCC(=O)OCC
Infrared spectrum
Conforms
GC
>=98.0 %
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text