Methyl caprate, 98+%, Thermo Scientific Chemicals
Methyl caprate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl caprate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 385550250
Price (USD)/ Each
25.65
Online exclusive
28.50 
Save 2.85 (10%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
25.65
Online exclusive
28.50 
Save 2.85 (10%)
Add to cart
Methyl caprate, 98+%, Thermo Scientific Chemicals
Catalog number385550250
Price (USD)/ Each
25.65
Online exclusive
28.50 
Save 2.85 (10%)
-
Add to cart
Chemical Identifiers
CAS110-42-9
IUPAC Namemethyl decanoate
Molecular FormulaC11H22O2
InChI KeyYRHYCMZPEVDGFQ-UHFFFAOYSA-N
SMILESCCCCCCCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.0 %
Appearance (Color)Clear colorless
Refractive index1.4240 to 1.4270 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only