Thermo Scientific Chemicals

Methyl nonanoate, 95%, Thermo Scientific Chemicals

Catalog number: 385560050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl nonanoate, 95%, Thermo Scientific Chemicals

Catalog number: 385560050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 385560050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1731-84-6
IUPAC Name
methyl nonanoate
Molecular Formula
C10H20O2
InChI Key
IJXHLVMUNBOGRR-UHFFFAOYSA-N
SMILES
CCCCCCCCC(=O)OC
Appearance (Form)
Liquid
Refractive index
1.4210 to 1.4230 (20°C, 589 nm)
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
GC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text