4-Phenyl-3-buten-2-one, 98+%
4-Phenyl-3-buten-2-one, 98+%
4-Phenyl-3-buten-2-one, 98+%
4-Phenyl-3-buten-2-one, 98+%
Thermo Scientific Chemicals

4-Phenyl-3-buten-2-one, 98+%

CAS: 122-57-6 | C10H10O | 146.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number 385592500
Price (USD)
30.65
Special offer
Online exclusive
Ends: 26-Sep-2025
37.80
Save 7.15 (19%)
Each
In stock
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
30.65
Special offer
Online exclusive
Ends: 26-Sep-2025
37.80
Save 7.15 (19%)
Each
Add to cart
Chemical Identifiers
CAS122-57-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting solid
Appearance (Color)Yellow
Infrared spectrumConforms
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection