Thermo Scientific Chemicals

Citric acid monohydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 385852500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Citric acid monohydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 385852500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 385852500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5949-29-1
IUPAC Name
2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
Molecular Formula
C6H10O8
InChI Key
YASYEJJMZJALEJ-UHFFFAOYSA-N
SMILES
O.OC(=O)CC(O)(CC(O)=O)C(O)=O
Appearance (Color)
Colorless to white
Phosphate (PO4)
=<0.001 %
Sulfate (SO4)
=<0.002 %
Appearance (Form)
Powder or crystals
Titration with NaOH
99.0 to 102.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text