Thermo Scientific Chemicals

4-Acetoxy-3-methoxybenzaldehyde, 96%, Thermo Scientific Chemicals

Catalog number: 385940250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Acetoxy-3-methoxybenzaldehyde, 96%, Thermo Scientific Chemicals

Catalog number: 385940250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 385940250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
881-68-5
IUPAC Name
4-formyl-2-methoxyphenyl acetate
Molecular Formula
C10H10O4
InChI Key
PZSJOBKRSVRODF-UHFFFAOYSA-N
SMILES
COC1=CC(C=O)=CC=C1OC(C)=O
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
73°C to 80°C
GC
>=97.5 %
Appearance (Color)
White to beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text