L(+)-Glutamine, Thermo Scientific Chemicals
L(+)-Glutamine, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L(+)-Glutamine, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
250 g
Catalog number 386030010
Price (USD)/ Each
-
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS56-85-9
IUPAC Name(2S)-2-amino-4-carbamoylbutanoic acid
Molecular FormulaC5H10N2O3
InChI KeyZDXPYRJPNDTMRX-VKHMYHEASA-N
SMILESN[C@@H](CCC(N)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO498.5 to 101.5 % (On dried basis)
Appearance (Form)Crystalline powder or flakes
Infrared spectrumConforms
Loss on drying=<0.3 % (1 g, 105°C, 3 h)
Residue after ignition=<0.3 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only