L(+)-Glutamine, specified according to the requirements of USP
L(+)-Glutamine, specified according to the requirements of USP
L(+)-Glutamine, specified according to the requirements of USP
L(+)-Glutamine, specified according to the requirements of USP
Thermo Scientific Chemicals

L(+)-Glutamine, specified according to the requirements of USP

CAS: 56-85-9 | C5H10N2O3 | 146.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 386030500
Price (USD)
53.65
Online Exclusive
59.40
Save 5.75 (10%)
Each
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
53.65
Online Exclusive
59.40
Save 5.75 (10%)
Each
Add to cart
Chemical Identifiers
CAS56-85-9
IUPAC Name(2S)-2-amino-4-carbamoylbutanoic acid
Molecular FormulaC5H10N2O3
InChI KeyZDXPYRJPNDTMRX-VKHMYHEASA-N
SMILESN[C@@H](CCC(N)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<0.3 % (1 g, 105°C, 3 h)
Titration with HClO498.5 to 101.5 % (On dried basis)
Appearance (Form)Crystalline powder or flakes
Infrared spectrumConforms
Residue after ignition=<0.3 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link