Sodium perborate monohydrate, 95%, Thermo Scientific Chemicals
Sodium perborate monohydrate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium perborate monohydrate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
250 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 386410010
Price (USD)/ Each
217.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
256.00 
Save 38.35 (15%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
217.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
256.00 
Save 38.35 (15%)
Add to cart
Sodium perborate monohydrate, 95%, Thermo Scientific Chemicals
Catalog number386410010
Price (USD)/ Each
217.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
256.00 
Save 38.35 (15%)
-
Add to cart
Chemical Identifiers
CAS10332-33-9
IUPAC Namesodium hydrate [(oxoboranyl)oxy]olate
Molecular FormulaBH2NaO4
InChI KeyMDGXUEVTGARGDK-UHFFFAOYSA-M
SMILESO.[Na+].[O-]OB=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline or granular powder
Titration with KMnO415 to 16 % (active oxygen)

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only